1,1,3,3-Tetramethylbutyl isocyanide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 1,1,3,3-Tetramethylbutylisocyanide

OR927941-25G OR927941-5G
APOSOR927941-25GEA 808 EUR
APOSOR927941-25G APOSOR927941-5G
1,1,3,3-Tetramethylbutyl isocyanide 95%
1,1,3,3-Tetramethylbutyl isocyanide
Formule: C₉H₁₇N
MW: 139.2380 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000003
CAS nummer: 14542-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR