4-Ethyl-1H-pyrrole-3-carboxylic acid methyl ester 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR928304-1G OR928304-250MG
APOSOR928304-1GEA 811 EUR
APOSOR928304-1G APOSOR928304-250MG
4-Ethyl-1H-pyrrole-3-carboxylic acid methyl ester 95%
4-Ethyl-1H-pyrrole-3-carboxylic acid methyl ester
Formule: C₈H₁₁NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11875848
CAS nummer: 1260827-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR