1-Furan-3-yl-ethylamine 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR929031-1G OR929031-250MG
APOSOR929031-1GEA 479 EUR
APOSOR929031-1G APOSOR929031-250MG
1-Furan-3-yl-ethylamine 96%
1-Furan-3-yl-ethylamine
Formule: C₆H₉NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06738794
CAS nummer: 252372-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR