N,N-Dimethylglycine methyl ester hydrochloride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR929353-1G OR929353-5G
APOSOR929353-1GEA 256 EUR
APOSOR929353-1G APOSOR929353-5G
N,N-Dimethylglycine methyl ester hydrochloride 95%
N,N-Dimethylglycine methyl ester hydrochloride
Formule: C₅H₁₁NO₂·ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08062582
CAS nummer: 1954-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR