Isopropyl-4-hydroxybenzoate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR930029-25G OR930029-5G
APOSOR930029-25GEA 271 EUR
APOSOR930029-25G APOSOR930029-5G
Isopropyl-4-hydroxybenzoate 95%
Isopropyl-4-hydroxybenzoate
Formule: C₁₀H₁₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016468
CAS nummer: 4191-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR