n-Propyl carbamate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR931217-100MG OR931217-1G OR931217-250MG OR931217-5G OR931217-25G
APOSOR931217-100MGEA 0 EUR
APOSOR931217-100MG APOSOR931217-1G APOSOR931217-250MG APOSOR931217-5G APOSOR931217-25G
n-Propyl carbamate 95%
n-Propyl carbamate
Formule: C₄H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00047879
CAS nummer: 627-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR