2-Bromoterephthalic acid 98+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR9323-100G OR9323-25G OR9323-5G
APOSOR9323-100GEA 139 EUR
APOSOR9323-100G APOSOR9323-25G APOSOR9323-5G
2-Bromoterephthalic acid 98+%
2-Bromoterephthalic acid
Formule: C₈H₅BrO₄
Smeltpunt: 295…297 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002403
CAS nummer: 586-35-6
EINECS: 209-572-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR