S-Ethyl thioacetate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR936522-100G OR936522-25G
APOSOR936522-100GEA 292 EUR
APOSOR936522-100G APOSOR936522-25G
S-Ethyl thioacetate 98%
S-Ethyl thioacetate
Formule: C₄H₈OS
MW: 104.17 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015178
CAS nummer: 625-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR