2-Chloro-N,N-diethylaniline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR936656-10G OR936656-1G OR936656-5G
APOSOR936656-10GEA 796 EUR
APOSOR936656-10G APOSOR936656-1G APOSOR936656-5G
2-Chloro-N,N-diethylaniline 95%
2-Chloro-N,N-diethylaniline
Formule: C₁₀H₁₄ClN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00059477
CAS nummer: 19372-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR