4-Chloro-(3-methoxycarbonyl)phenylboronic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4-Chloro-3-(methoxycarbonyl)benzeneboronic acid, 4-Chloro-3-(methoxycarbonyl)phenylboronic acid

OR9368-1G OR9368-5G OR9368-10G
APOSOR9368-1GEA 22 EUR
APOSOR9368-1G APOSOR9368-5G APOSOR9368-10G
4-Chloro-(3-methoxycarbonyl)phenylboronic acid 98%
4-Chloro-(3-methoxycarbonyl)phenylboronic acid
Formule: C₈H₈BClO₄
Smeltpunt: 136…138 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06801690
CAS nummer: 874219-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR