4-Octanol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR937548-25G OR937548-5G
APOSOR937548-25GEA 482 EUR
APOSOR937548-25G APOSOR937548-5G
4-Octanol 97%
4-Octanol
Formule: C₈H₁₈O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014409
CAS nummer: 589-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR