Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR937721-1G OR937721-250MG
APOSOR937721-1GEA 232 EUR
APOSOR937721-1G APOSOR937721-250MG
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate 95%
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate
Formule: C₁₁H₁₅NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06227773
CAS nummer: 65880-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR