3-Methylbenzenesulphonamide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR938766-1G OR938766-250MG
APOSOR938766-1GEA 226 EUR
APOSOR938766-1G APOSOR938766-250MG
3-Methylbenzenesulphonamide 95%
3-Methylbenzenesulphonamide
Formule: C₇H₉NO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04066695
CAS nummer: 1899-94-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR