5-Bromo-2-methoxybenzenesulphonamide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR9399-1G OR9399-250MG OR9399-5G
APOSOR9399-1GEA 141 EUR
APOSOR9399-1G APOSOR9399-250MG APOSOR9399-5G
5-Bromo-2-methoxybenzenesulphonamide 95%
5-Bromo-2-methoxybenzenesulphonamide
Formule: C₇H₈BrNO₃S
Smeltpunt: 151…153 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06808831
CAS nummer: 23095-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR