3-(Cyclopropylmethoxy)-4-hydroxybenzaldehyde 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR940968-1G OR940968-250MG
APOSOR940968-1GEA 149 EUR
APOSOR940968-1G APOSOR940968-250MG
3-(Cyclopropylmethoxy)-4-hydroxybenzaldehyde 95%
3-(Cyclopropylmethoxy)-4-hydroxybenzaldehyde
Formule: C₁₁H₁₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16999806
CAS nummer: 25934-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR