N-Benzyl-5-bromo-2-methoxybenzenesulphonamide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR9413-1G OR9413-5G
APOSOR9413-1GEA 139 EUR
APOSOR9413-1G APOSOR9413-5G
N-Benzyl-5-bromo-2-methoxybenzenesulphonamide 97%
N-Benzyl-5-bromo-2-methoxybenzenesulphonamide
Formule: C₁₄H₁₄BrNO₃S
Smeltpunt: 136…140 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02227932
CAS nummer: 446308-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR