2-Methylisophthalic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR941367-1G
APOSOR941367-1GEA 395 EUR
APOSOR941367-1G
2-Methylisophthalic acid 95%
2-Methylisophthalic acid
Formule: C₉H₈O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00157035
CAS nummer: 15120-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR