2,6-Dimethyl-3-nitroaniline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR941380-1G OR941380-5G OR941380-25G
APOSOR941380-1GEA 45.4 EUR
APOSOR941380-1G APOSOR941380-5G APOSOR941380-25G
2,6-Dimethyl-3-nitroaniline 95%
2,6-Dimethyl-3-nitroaniline
Formule: C₈H₁₀N₂O₂
MW: 166.177 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00156348
CAS nummer: 67083-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR