6-tert-Butyl-o-cresol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR942617-25G OR942617-100G OR942617-500G
APOSOR942617-25GEA 132 EUR
APOSOR942617-25G APOSOR942617-100G APOSOR942617-500G
6-tert-Butyl-o-cresol 95%
6-tert-Butyl-o-cresol
Formule: C₁₁H₁₆O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002239
CAS nummer: 2219-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR