6-tert-Butyl-m-cresol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR950844-500G
APOSOR950844-500GEA 68.7 EUR
APOSOR950844-500G
6-tert-Butyl-m-cresol 95%
6-tert-Butyl-m-cresol
Formule: C₁₁H₁₆O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002308
CAS nummer: 88-60-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR