3-(2-Aminoethoxy)benzonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR951396-1G
APOSOR951396-1GEA 64.3 EUR
APOSOR951396-1G
3-(2-Aminoethoxy)benzonitrile 95%
3-(2-Aminoethoxy)benzonitrile
Formule: C₉H₁₀N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16495888
CAS nummer: 120351-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR