Pseudopelletierine oxime 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 9-methyl-9-azabicyclo[3.3.1]nonan-3-one oxime

OR951413-100G OR951413-25G
APOSOR951413-100GEA 233 EUR
APOSOR951413-100G APOSOR951413-25G
Pseudopelletierine oxime 98%
Pseudopelletierine oxime
Formule: C₉H₁₆N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03755545
CAS nummer: 6164-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR