3-Chloro-5-nitropyridin-4-ol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR952268-1G OR952268-250MG
APOSOR952268-1GEA 52.6 EUR
APOSOR952268-1G APOSOR952268-250MG
3-Chloro-5-nitropyridin-4-ol 95%
3-Chloro-5-nitropyridin-4-ol
Formule: C₅H₃ClN₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11044290
CAS nummer: 31872-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR