4,5-Dichloroquinazoline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR953494-1G OR953494-250MG
APOSOR953494-1GEA 127 EUR
APOSOR953494-1G APOSOR953494-250MG
4,5-Dichloroquinazoline 95%
4,5-Dichloroquinazoline
Formule: C₈H₄Cl₂N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09909637
CAS nummer: 2148-55-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR