4-[(Methylsulphanyl)methyl]benzoic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR954025-1G OR954025-250MG
APOSOR954025-1GEA 438 EUR
APOSOR954025-1G APOSOR954025-250MG
4-[(Methylsulphanyl)methyl]benzoic acid 98%
4-[(Methylsulphanyl)methyl]benzoic acid
Formule: C₉H₁₀O₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06373463
CAS nummer: 67003-48-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR