3-Methyl-5-phenylphenol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR954883-1G
APOSOR954883-1GEA 505 EUR
APOSOR954883-1G
3-Methyl-5-phenylphenol 98%
3-Methyl-5-phenylphenol
Formule: C₁₃H₁₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01869167
CAS nummer: 50715-82-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR