4-Chloro-2-methyl-benzoic acid methyl ester 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methyl 4-chloro-2-methylbenzoate

OR955773-5G
APOSOR955773-5GEA 145 EUR
APOSOR955773-5G
4-Chloro-2-methyl-benzoic acid methyl ester 98%
4-Chloro-2-methyl-benzoic acid methyl ester
Formule: C₉H₉ClO₂
MW: 184.62 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09842344
CAS nummer: 99585-12-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR