N-(4-Nitrobenzyl)acetamide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR956367-1G
APOSOR956367-1GEA 330 EUR
APOSOR956367-1G
N-(4-Nitrobenzyl)acetamide 95%
N-(4-Nitrobenzyl)acetamide
Formule: C₉H₁₀N₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00230940
CAS nummer: 56222-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR