4-Chloro-3,5-dimethylisoxazole 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR956927-1G
APOSOR956927-1GEA 490 EUR
APOSOR956927-1G
4-Chloro-3,5-dimethylisoxazole 95%
4-Chloro-3,5-dimethylisoxazole
Formule: C₅H₆ClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD14702892
CAS nummer: 10557-86-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR