4-Hydroxy-N-methylbenzene-1-sulphonamide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR957676-1G
APOSOR957676-1GEA 636 EUR
APOSOR957676-1G
4-Hydroxy-N-methylbenzene-1-sulphonamide 95%
4-Hydroxy-N-methylbenzene-1-sulphonamide
Formule: C₇H₉NO₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01707199
CAS nummer: 3077-61-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR