4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959405-1G OR959405-5G
APOSOR959405-1GEA 120 EUR
APOSOR959405-1G APOSOR959405-5G
4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile 98%
4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile
Formule: C₁₀H₁₁N₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08062860
CAS nummer: 127680-86-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR