Methyl 2-methoxy-5-methylpyridine-3-carboxylate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959608-1G OR959608-250MG
APOSOR959608-1GEA 636 EUR
APOSOR959608-1G APOSOR959608-250MG
Methyl 2-methoxy-5-methylpyridine-3-carboxylate 95%
Methyl 2-methoxy-5-methylpyridine-3-carboxylate
Formule: C₉H₁₁NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16611638
CAS nummer: 351410-41-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR