5-bromo-4-methoxypyrimidine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959616-1G OR959616-250MG
APOSOR959616-1GEA 158 EUR
APOSOR959616-1G APOSOR959616-250MG
5-bromo-4-methoxypyrimidine 95%
5-bromo-4-methoxypyrimidine
Formule: C₅H₅BrN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00234066
CAS nummer: 4319-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR