2-Chloro-5-methyl-4-nitropyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959694-1G OR959694-250MG
APOSOR959694-1GEA 52.6 EUR
APOSOR959694-1G APOSOR959694-250MG
2-Chloro-5-methyl-4-nitropyridine 95%
2-Chloro-5-methyl-4-nitropyridine
Formule: C₆H₅ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD19690032
CAS nummer: 97944-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR