tert-Butyl pyridine-2-carboxylate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR959821-1G OR959821-5G
APOSOR959821-1GEA 220 EUR
APOSOR959821-1G APOSOR959821-5G
tert-Butyl pyridine-2-carboxylate 98%
tert-Butyl pyridine-2-carboxylate
Formule: C₁₀H₁₃NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD19600064
CAS nummer: 79233-72-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR