3-(Cyclopropylmethoxy)azetidine hydrochloride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR960080-1G OR960080-5G
APOSOR960080-1GEA 292 EUR
APOSOR960080-1G APOSOR960080-5G
3-(Cyclopropylmethoxy)azetidine hydrochloride 95%
3-(Cyclopropylmethoxy)azetidine hydrochloride
Formule: C₇H₁₃NO·ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22205833
CAS nummer: 1375069-07-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR