4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR960181-5G
APOSOR960181-5GEA 210 EUR
APOSOR960181-5G
4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid 97%
4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid
Formule: C₉H₁₀O₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00652575
CAS nummer: 19156-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR