3-(tert-Butyl)-5-methyl-1H-pyrazole 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR960601-1G
APOSOR960601-1GEA 330 EUR
APOSOR960601-1G
3-(tert-Butyl)-5-methyl-1H-pyrazole 97%
3-(tert-Butyl)-5-methyl-1H-pyrazole
Formule: C₈H₁₄N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18325158
CAS nummer: 96440-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR