1-Boc-4-bromopiperidine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR962067-100G OR962067-25G OR962067-5G OR962067-1G
APOSOR962067-100GEA 0 EUR
APOSOR962067-100G APOSOR962067-25G APOSOR962067-5G APOSOR962067-1G
1-Boc-4-bromopiperidine 97%
1-Boc-4-bromopiperidine
Formule: C₁₀H₁₈BrNO₂
Kookpunt: 100…118 °C/0,5mmHg
Dichtheid: 1,3403 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115039
CAS nummer: 180695-79-8
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR