(R)-3-(Fmoc-amino)-3-phenylpropionic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR962068-1G OR962068-5G
APOSOR962068-1GEA 80.4 EUR
APOSOR962068-1G APOSOR962068-5G
(R)-3-(Fmoc-amino)-3-phenylpropionic acid 97%
(R)-3-(Fmoc-amino)-3-phenylpropionic acid
Formule: C₂₄H₂₁NO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01311783
CAS nummer: 220498-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR