Fmoc-Dab(Alloc)-OH 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR962282-1G OR962282-25G OR962282-5G
APOSOR962282-1GEA 77.5 EUR
APOSOR962282-1G APOSOR962282-25G APOSOR962282-5G
Fmoc-Dab(Alloc)-OH 98%
Fmoc-Dab(Alloc)-OH
Formule: C₂₃H₂₄N₂O₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00798634
CAS nummer: 204316-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR