4-Benzyloxypiperidine hydrochloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR962669-1G OR962669-5G
APOSOR962669-1GEA 193 EUR
APOSOR962669-1G APOSOR962669-5G
4-Benzyloxypiperidine hydrochloride 97%
4-Benzyloxypiperidine hydrochloride
Formule: C₁₂H₁₈ClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07368306
CAS nummer: 81151-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR