4-(Pyrrolidin-1-yl)benzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR962672-25G OR962672-5G
APOSOR962672-25GEA 238 EUR
APOSOR962672-25G APOSOR962672-5G
4-(Pyrrolidin-1-yl)benzonitrile 98%
4-(Pyrrolidin-1-yl)benzonitrile
Formule: C₁₁H₁₂N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07368512
CAS nummer: 10282-30-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR