2-(1-Pyrrolidinyl)benzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR962734-1G OR962734-5G
APOSOR962734-1GEA 77.5 EUR
APOSOR962734-1G APOSOR962734-5G
2-(1-Pyrrolidinyl)benzoic acid 97%
2-(1-Pyrrolidinyl)benzoic acid
Formule: C₁₁H₁₃NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09035985
CAS nummer: 78648-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR