(4-Bromothiophen-2-yl)methanamine hydrobromide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR962877-1G OR962877-250MG
APOSOR962877-1GEA 307 EUR
APOSOR962877-1G APOSOR962877-250MG
(4-Bromothiophen-2-yl)methanamine hydrobromide 95%
(4-Bromothiophen-2-yl)methanamine hydrobromide
Formule: C₅H₆BrNS·BrH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11855809
CAS nummer: 1314707-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR