2-Bromo-4-chlorotoluene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR9677-100G
APOSOR9677-100GEA 55.5 EUR
APOSOR9677-100G
2-Bromo-4-chlorotoluene
2-Bromo-4-chlorotoluene
Formule: C₇H₆BrCl
Kookpunt: 114…12mm °C
Dichtheid: 156 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00060650
CAS nummer: 27139-97-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR