3-Chlorothioanisole 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (3-Chlorophenyl)(methyl)sulphane, 3-Chlorophenyl methyl sulphide, 1-Chloro-3-(methylthio)benzene

OR9711-100G OR9711-25G OR9711-5G
APOSOR9711-100GEA 366 EUR
APOSOR9711-100G APOSOR9711-25G APOSOR9711-5G
3-Chlorothioanisole 97%
3-Chlorothioanisole
Formule: C₇H₇ClS
MW: 158.65 g/mol
Kookpunt: 218 °C
Dichtheid: 1.206 g/cm³ (25 °C)
Vlampunt: 106 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00040127
CAS nummer: 4867-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR