4-Hydroxy-3-iodobenzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR9769-250MG
APOSOR9769-250MGEA 65.8 EUR
APOSOR9769-250MG
4-Hydroxy-3-iodobenzoic acid 97%
4-Hydroxy-3-iodobenzoic acid
Formule: C₇H₅IO₃
Smeltpunt: 170…175 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00234263
CAS nummer: 37470-46-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR