2,4-Dimethylpyrrole 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR9930-5G OR9930-25G
APOSOR9930-5GEA 73 EUR
APOSOR9930-5G APOSOR9930-25G
2,4-Dimethylpyrrole 98%
2,4-Dimethylpyrrole
Formule: C₆H₉N
Kookpunt: 165…167 °C
Dichtheid: 0,924 g/cm³ (25 °C)
Vlampunt: >113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00192088
CAS nummer: 625-82-1
EINECS: 210-912-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR