Cyclopropyl(trimethylsilyl)acetylene 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR9948-5G
APOSOR9948-5GEA 116 EUR
APOSOR9948-5G
Cyclopropyl(trimethylsilyl)acetylene 96%
Cyclopropyl(trimethylsilyl)acetylene
Formule: C₈H₁₄Si
Kookpunt: 83 °C/80mmHg
Dichtheid: 1,2726 g/cm³ (25 °C)
Vlampunt: 25 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00671355
CAS nummer: 81166-84-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR